Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBPF15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583644 Supplier Invitrogen™ Supplier No. PA583644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGCLELTDSC QPYRSAFYVL EQQRV Highest antigen sequence indentity to the following orthologs: Mouse - 40%, Rat - 40%.

NBPF15 (NBPF Member 15), located on 1q21.1, is a Protein Coding gene. This gene is a member of the neuroblastoma breakpoint family (NBPF) which consists of dozens of recently duplicated genes primarily located in segmental duplications on human chromosome 1. This gene family contains numerous pseudogenes. Human NBPF15 encodes a 670 AA protein consisting of six clades of Olduvai domains. The entanglements between the mosaic disorder-ordered segments in the NBPF15 N terminus have triggered the multiple types of aggregates and phase transition of NBPF15 proteins, which could be associated with Olduvai-related cognitive dysfunction diseases. Diseases associated with NBPF15 include Neuroblastoma and Hydrolethalus Syndrome 1. An important paralog of this gene is NBPF14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBPF15
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NBPF15
Gene Accession No. Q8N660
Gene Alias AB14; NBPF; NBPF Member 15; Neuroblastoma Breakpoint Family, Member 1; Neuroblastoma Breakpoint Family, Member 16
Gene Symbols NBPF15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NBPF15. Recombinant protein control fragment (Product #RP-108172).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284565
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.