Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBPF Pan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101712 Supplier Invitrogen™ Supplier No. PA5101712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NBPF1/9/10/12/14/15/16/20.

This gene is a member of the neuroblastoma breakpoint family (NBPF) which consists of dozens of recently duplicated genes primarily located in segmental duplications on human chromosome 1. This gene family has experienced its greatest expansion within the human lineage and has expanded, to a lesser extent, among primates in general. Members of this gene family are characterized by tandemly repeated copies of DUF1220 protein domains. Gene copy number variations in the human chromosomal region 1q21.1, where most DUF1220 domains are located, have been implicated in a number of developmental and neurogenetic diseases such as microcephaly, macrocephaly, autism, schizophrenia, mental retardation, congenital heart disease, neuroblastoma, and congenital kidney and urinary tract anomalies. Altered expression of some gene family members is associated with several types of cancer. This gene family contains numerous pseudogenes. [provided by RefSeq, Apr 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBPF Pan
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NBPF1
Gene Accession No. P0DPF2, P0DPF3, Q3BBV0, Q5TAG4, Q5TI25, Q6P3W6, Q8N660
Gene Alias AB13; AB14; AB23; AB6; AD2; AE01; AE5; AG1; AG3; chomosome one amplified sequence 1 cyclophilin; chromosome 1 amplified sequence 1; COAS1; DJ328E19.C1.1; KIAA1245; KIAA1693; NBFP10; NBFP12; NBFP14; NBFP15; NBFP16; NBFP20; NBFP9; NBG; NBPF; NBPF1; NBPF10; NBPF12; NBPF14; NBPF15; NBPF16; NBPF20; NBPF9; Neuroblastoma breakpoint family member 1; neuroblastoma breakpoint family member 10; neuroblastoma breakpoint family member 12; neuroblastoma breakpoint family member 14; neuroblastoma breakpoint family member 15; Neuroblastoma breakpoint family member 16; neuroblastoma breakpoint family member 20; Neuroblastoma breakpoint family member 9; neuroblastoma breakpoint family, member 1; neuroblastoma breakpoint family, member 10; neuroblastoma breakpoint family, member 12; neuroblastoma breakpoint family, member 14; neuroblastoma breakpoint family, member 15; neuroblastoma breakpoint family, member 16; neuroblastoma breakpoint family, member 20; neuroblastoma breakpoint family, member 9
Gene Symbols NBPF1, NBPF10, NBPF12, NBPF14, NBPF15, NBPF20, NBPF9
Host Species Rabbit
Immunogen A synthesized peptide derived from human NBPF1(Accession Q3BBV0), corresponding to amino acid residues L1075-E1125.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100132406, 100288142, 149013, 25832, 284565, 400818, 55672
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.