Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NBPF11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558527
Description
Immunogen sequence: EEDKVNSSLV VDRESSHDGC QDALN Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 40%.
NBPF11 is a protein coding gene.
Specifications
NBPF11 | |
Polyclonal | |
Unconjugated | |
NBPF11 | |
NBPF11; NBPF24; Neuroblastoma breakpoint family member 11; neuroblastoma breakpoint family member 24; neuroblastoma breakpoint family, member 11; neuroblastoma breakpoint family, member 24 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200030 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86T75 | |
NBPF11 | |
Recombinant protein corresponding to Human NBPF11. Recombinant protein control fragment (Product #RP-104314). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction