Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NBPF15 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306172UV
Description
NBPF15 Polyclonal specifically detects NBPF15 in Human, Cynomolgus Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NBPF15 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
AB14, Ag3, NBPF16, Neuroblastoma Breakpoint Family Member 15, Neuroblastoma Breakpoint Family Member 16, Neuroblastoma Breakpoint Family, Member 15, Neuroblastoma Breakpoint Family, Member 16 | |
Produced in rabbits immunized with E. coli-derived Human NBPF15 fragment. | |
0.1 mL | |
Primary | |
Human, Cynomolgus Monkey | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
284565 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction