Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBPF5P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101713 Supplier Invitrogen™ Supplier No. PA5101713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NBPF5.

NBPF5P is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBPF5P
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NBPF5P
Gene Accession No. Q86XG9
Gene Alias NBPF5; NBPF5P
Gene Symbols NBPF5P
Host Species Rabbit
Immunogen A synthesized peptide derived from human NBPF5P(Accession Q86XG9), corresponding to amino acid residues A305-N351.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100507044
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.