Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBPF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103962 Supplier Invitrogen™ Supplier No. PA5103962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NBPF6.

Members of the neuroblastoma breakpoint family are encoded by genes that map to a region of segmental duplications on human chromosome 1. Both NBPF4 (neuroblastoma breakpoint family member 4) and NBPF6 (neuroblastoma breakpoint family member 6) are cytoplasmic proteins that belong to the NBPF family. NBPF4 contains 123 amino acids and is expressed in testis, while NBPF6 consists of 638 amino acids and contains 3 NBPF domains. Like other NBPF family members, both proteins are encoded by genes that map to human chromosome 1p13.3. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBPF6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NBPF6
Gene Accession No. Q5VWK0
Gene Alias NBPF6; Neuroblastoma breakpoint family member 6; neuroblastoma breakpoint family, member 6
Gene Symbols NBPF6
Host Species Rabbit
Immunogen A synthesized peptide derived from human NBPF6(Accession Q5VWK0), corresponding to amino acid residues E505-L555.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653149
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.