Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBR1 Mouse anti-Human, Unconjugated, Clone: 5C3, Abnova™

Catalog No. 89339159 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-159 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-159 Supplier Abnova Corporation Supplier No. H00004077M05J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

NBR1 Monoclonal antibody specifically detects NBR1 in Human samples. It is validated for S-ELISA, ELISA, WB-Re, WB-Tr, RNAi-Ab.

Specifications

Antigen NBR1
Applications ELISA, Western Blot, KnockDown
Classification Monoclonal
Clone 5C3
Conjugate Unconjugated
Description neighbor of BRCA1 gene 1
Gene NBR1
Gene Accession No. NM_005899
Gene Alias 1A1-3B, KIAA0049, M17S2, MIG19
Host Species Mouse
Immunogen NBR1 (NP_005890, 2 a.a. ∼ 96 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 4077
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.