Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554946 Supplier Invitrogen™ Supplier No. PA554946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLHEKLVLQN PSLGSCPSEV SMPTSEETLF LPENQFSWHI ACNNCQRRIV GVRYQCSLCP SYNICEDCEA GPYGHDTNHV LLKLRRPVVG SSEPFCHS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 82%.

The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NBR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NBR1
Gene Accession No. Q14596
Gene Alias 1A13B; 1A1-3B; B-box protein; Ca125; Cell migration-inducing gene 19 protein; IAI3B; KIAA0049; M17S2; Membrane component chromosome 17 surface marker 2; Membrane component chromosome 17 surface marker 2 homolog; membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigen CA125); MIG19; migration-inducing protein 19; mKIAA0049; Nbr1; NBR1, autophagy cargo receptor; neighbor of Brca1 gene 1; neighbor of BRCA1 gene 1 protein; Next to BRCA1 gene 1 protein; next to the Brca1; ovarian carcinoma antigen CA125; Protein 1A1-3B; RGD1311421
Gene Symbols NBR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NBR1. Recombinant protein control fragment (Product #RP-93533).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.