Learn More
NBR1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305272A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene.Specifications
NBR1 | |
Polyclonal | |
Unconjugated | |
NBR1 | |
1A1-3B, M17S2, cell migration-inducing gene 19 protein, membrane component chromosome 17 surface marker 2, membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigen CA125), migration-inducing protein 19, neighbor of BRCA1 gene 1 protein, next to BRCA1 ge | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4077 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q14596 | |
NBR1 | |
Between 916 and 966 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.