Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nbs1 Antibody (7E4A2), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen Nbs1
Clone 7E4A2
Concentration 1.0 mg/mL
Dilution Western Blot 1:500 - 1:2000, Simple Western 1:50, Flow Cytometry 1:200 - 1:400, ELISA, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1-50 - 1:100, Immunohistochemistry-Paraffin, Knockout Validated
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP226297SS
SDP
View Documents
Novus Biologicals
NBP226297SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP226297
SDP
View Documents
Novus Biologicals
NBP226297
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Nbs1 Monoclonal specifically detects Nbs1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications

Specifications

Nbs1
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Unconjugated
Mouse
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
O60934
4683
Nbs1 Antibody (7E4A2) was made to partial recombinant human NBS1 (between residues 400-650) expressed in E. coli [Uniprot O60934]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
7E4A2
Western Blot 1:500 - 1:2000, Simple Western 1:50, Flow Cytometry 1:200 - 1:400, ELISA, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1-50 - 1:100, Immunohistochemistry-Paraffin, Knockout Validated
Monoclonal
Precipitation
RUO
Human
ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex
NBN
IgG2 α
Ammonium sulfate precipitation
85 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.