Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nbs1, p Ser343 Antibody (SR2302), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Nbs1 |
---|---|
Clone | SR2302 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
Nbs1 Monoclonal antibody specifically detects Nbs1 in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
Nbs1 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
A synthesized peptide derived from human phospho-Nbs1 [Ser343] (Uniprot #: O60934) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2302 | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4683 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title