Learn More
Invitrogen™ NBS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121126
Description
Positive test controls include: Jurkat, HeLa, MCF7, Mouse heart, Rat kidney. The target is usually found in the following locations: Chromosome, Nucleus, PML body, telomere. Immunogen sequence: LPKKLLLTEF RSLVIKNSTS RNPSGINDDY GQLKNFKKFK KVTYPGAGKL PHIIGGSDLI AHHARKNTEL EEWLRQEMEV QNQHAKEESL ADDLFRYNPY L.
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
Specifications
| NBS1 | |
| Polyclonal | |
| Unconjugated | |
| NBN | |
| AT V1; AT V2; ATV; AT-V1; AT-V2; Cell cycle regulatory protein p95; FLJ10155; I79_012607; MGC87362; Nbn; NBS; NBS1; nibrin; Nijmegen breakage syndrome 1 (nibrin); Nijmegen breakage syndrome protein 1; Nijmegen breakage syndrome protein 1 homolog; OTTHUMP00000204088; P95; p95 protein of the MRE11/RAD50 complex; p95-NBS1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27354, 4683, 85482 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60934, Q9JIL9, Q9R207 | |
| NBN | |
| A synthetic peptide corresponding to a sequence within amino acids 650-750 of human NBS1/NBS1/NBN (NP_0024762). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.