Learn More
NBS1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300187A
Description
The recommended shelf life for this product is 1 year from date of receipt.
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.Specifications
| NBS1 | |
| Polyclonal | |
| Unconjugated | |
| NBN | |
| AT-V1, AT-V2, ATV, FLJ10155, MGC87362, NBS, NBS1, P95 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4683 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O60934 | |
| NBN | |
| Between 625 and 675 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.