Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCAM-1/CD56 Antibody (301040), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2408M
This item is not returnable.
View return policy
Description
NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA (Matched Antibody Pair)Specifications
| NCAM-1/CD56 | |
| Monoclonal | |
| DyLight 594 | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| Mouse myeloma cell line NS0-derived recombinant human NCAM-1/CD56 | |
| 0.5 mL | |
| Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 4684 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA | |
| 301040 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction