Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM-1 (CD56) Mouse anti-Human, Brilliant Violet 510, Clone: R19-760, BD Optibuild™

Catalog No. BDB744218 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744218 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744218 Supplier BD Biosciences Supplier No. 744218
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The R19-760 monoclonal antibody specifically binds to NCAM-1 (Neural cell adhesion molecule 1) that is also known as CD56 and NKH1. NCAM-1 is a heavily glycosylated transmembrane protein and a member of the Ig supergene family. NCAM-1 represents an isoform of the neural cell adhesion molecule (NCAM). In the hematopoietic system, it is present on approximately 10% to 25% of peripheral blood lymphocytes. It is expressed on natural killer (NK) cells and a subset of T cells, NKT cells. It is not expressed on myeloid cells, erythrocytes or B cells. This molecule appears to function as an adhesion molecule and can serve as a panspecific NK-cell marker. The R19-760 antibody recognizes an epitope in the NCAM-1 (CD56) extracellular domain that resists destruction due to cellular fixation with BD Phosflow™ Lyse/Fix Buffer and permeabilization with BD Phosflow™ Perm Buffer III.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAM-1 (CD56)
Classification Monoclonal
Clone R19-760
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias N-CAM-1, Neural cell adhesion molecule 1, NKH1, MSK39, Leu-19
Host Species Mouse
Immunogen Human NCAM1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.