Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCAM-1/CD56 Antibody (735), Alexa Fluor™ 350 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252669AF350
Description
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
NCAM-1/CD56 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
NCAM1 | |
Homopolymers of alpha 2,8-linked sialic acid of Escherichia coli K1. | |
Primary | |
Recognises Polysialic acid (PSA), a carbohydrate polymer attached to the neural cell adhesion molecule (NCAM), which shares molecular epitopes with homopolymers of (alpha)2,8-linked sialic acid of Escherichia coli K1. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
735 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
Rabbit | |
0.1 mL | |
4684 | |
Human, Mouse, Rat | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction