Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCAM1/CD56 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E8C9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NCAM1/CD56 |
---|---|
Clone | 1E8C9 |
Concentration | 0.95 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6338
![]() ![]() |
Proteintech Group Inc
602381IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
NCAM, as a member of the immunoglobulin superfamily of adhesion molecules is characterized by several immunoglobulin (Ig)-like domains. The extracellular part of NCAM consists of five of these Ig domains and two fibronectin type III homology regions. NCAM is encoded by a single copy gene composed of 26 exons. However, at least 20-30 distinct isoforms can be generated by alternative splicing and by posttranslational modifications, such as sialylation. During sialylation, polysialic acid (PSA) carbohydrates are attached to the extracellular part of NCAM. Through its extracellular region, NCAM mediates homophilic interactions. In addition, NCAM can also undergo heterophilic interactions by binding extracellular matrix components, such as laminin, or other cell adhesion molecules, such as integrins.Specifications
NCAM1/CD56 | |
0.95 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD56, MSK39, N CAM 1, NCAM, NCAM 1, NCAM1 | |
Ncam1 | |
IgG1 | |
Protein G | |
Antibody |
1E8C9 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
P13591, P13595, P13596 | |
100515564, 17967, 24586, 4684 | |
NCAM1/CD56 Fusion Protein Ag5732 | |
Primary | |
-20°C | |
NCAM1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title