Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM1 Mouse anti-Human, Clone: NCAM1/1496, Abnova™

Catalog No. 89034774 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-774 Supplier Abnova Corporation Supplier No. MAB14691

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against recombinant human NCAM1.

  • High specificity
  • High reactivity

Specifications

Antigen NCAM1
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone NCAM1/1496
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human NCAM1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA and 0.05% azide).
Gene Alias CD56/MSK39/NCAM
Gene Symbols NCAM1
Host Species Mouse
Immunogen Recombinant protein corresponding to human NCAM1.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4684
Test Specificity Reacts with an extracellular domain (close to transmembrane) of CD56/NCAM.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.