Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM1 Mouse anti-Human, PE, Clone: COC56, Abnova™

Catalog No. 89934655 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-655 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-655 Supplier Abnova Corporation Supplier No. MAB12410
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human NCAM1.

Specifications

Antigen NCAM1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone COC56
Conjugate PE
Description Mouse monoclonal antibody raised against human NCAM1.
Dilution Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
Gene Alias CD56/MSK39/NCAM
Gene Symbols NCAM1
Host Species Mouse
Immunogen NCAM1
Purification Method Protein A/G affinity chromatography
Quantity 50 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4684
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.