Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556584
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556584 Supplier Invitrogen™ Supplier No. PA556584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVPPAISMPQ KSFNATAERG EEMTFSCRAS GSPEPAISWF RNGKLIEENE KYILKGSNTE LTVRNIINSD GGPY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAM2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCAM2
Gene Accession No. O15394, O35136
Gene Alias fasciclin II; NCAM2; N-CAM-2; Ncam-2; NCAM21; neural cell adhesion molecule 2; Neural cell adhesion molecule RB-8; Ocam; OCAM-GPI; olfactory axon cell adhesion molecule; R4B12; R4B12 antigen; Rncam
Gene Symbols NCAM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NCAM2. Recombinant protein control fragment (Product #RP-95877).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17968, 4685
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.