Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110015 Supplier Invitrogen™ Supplier No. PA5110015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YGVKIIELSQ TTAKVSFNKP DSHGGVPIHH YQVDVKEVAS EIWKIVRSHG VQTMVVLNNL EPNTTYEIRV AAVNGKGQGD YSKIEIFQTL PVREPSPPSI HGQPSSGKSF KLSITKQDDG GAPILEYIVK YRSKDKEDQW LEKKVQGNKD HIILEHLQWT MGYEVQITAA NRLGYSEPTV YEFSMPPKPN I.

NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAM2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NCAM2
Gene Accession No. O15394, O35136
Gene Alias fasciclin II; NCAM2; N-CAM-2; Ncam-2; NCAM21; neural cell adhesion molecule 2; Neural cell adhesion molecule RB-8; Ocam; OCAM-GPI; olfactory axon cell adhesion molecule; R4B12; R4B12 antigen; Rncam
Gene Symbols NCAM2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 500-690 of human NCAM2 (NP_004531.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17968, 288280, 4685
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.