Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCAPD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110016 Supplier Invitrogen™ Supplier No. PA5110016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGDKLSDESI FSAFLSVVGK LRRGAKPEGK AIIDEFEQKL RACHTRGLDG IKELEIGQAG SQRAPSAKKP STGSRYQPLA STASDNDFVT PEPRRTTRRH P.

Regulatory subunit of the condensin complex, a complex required for conversion of interphase chromatin into mitotic-like condense chromosomes. The condensin complex probably introduces positive supercoils into relaxed DNA in the presence of type I topoisomerases and converts nicked DNA into positive knotted forms in the presence of type II topoisomerases. May target the condensin complex to DNA via its C-terminal domain (PubMed:11136719). May promote the resolution of double-strand DNA catenanes (intertwines) between sister chromatids. Condensin-mediated compaction likely increases tension in catenated sister chromatids, providing directionality for type II topoisomerase-mediated strand exchanges toward chromatid decatenation. Required for decatenation of non-centromeric ultrafine DNA bridges during anaphase. Early in neurogenesis, may play an essential role to ensure accurate mitotic chromosome condensation in neuron stem cells, ultimately affecting neuron pool and cortex size (PubMed:27737959). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAPD2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NCAPD2
Gene Accession No. Q15021, Q8K2Z4
Gene Alias 2810406C15Rik; 2810465G24Rik; Capd2; CAP-D2; chromosome condensation related SMC associated protein 1; chromosome condensation-related SMC-associated protein 1; Chromosome-associated protein D2; CNAP1; Condensin complex subunit 1; hCAP-D2; KIAA0159; mCAP-D2; mKIAA0159; Ncapd2; Non-SMC condensin I complex subunit D2; non-SMC condensin I complex, subunit D2; RGD1562596; XCAP-D2 homolog
Gene Symbols NCAPD2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1250-1350 of human NCAPD2 (NP_0556803).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362438, 68298, 9918
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.