Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCAPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565262 Supplier Invitrogen™ Supplier No. PA565262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFHIWSKKEKF SPTFINNVIS HTGTEHSAPA WMLLSKIAGS SPRLDYSRII QSWEKISSQQ NPNSNTLGHI LCVIGHIAKH LPKSTRDKVT DAVKCK Highest antigen sequence identity to the following orthologs - mouse 75%, rat 75%.

This protein is a regulatory subunit of the condensin-2 complex, a complex which establishes mitotic chromosome architecture and is involved in physical rigidity of the chromatid axis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAPD3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCAPD3
Gene Accession No. P42695
Gene Alias 2810487N22Rik; 4632407J06Rik; AI195468; AU018739; B130055D15Rik; CAPD3; CAP-D3; Condensin-2 complex subunit D3; hCAP-D3; hcp-6; hHCP-6; Jacobsen syndrome region 2; Kiaa0056; mKIAA0056; NCAPD3; Non-SMC condensin II complex subunit D3; non-SMC condensin II complex, subunit D3
Gene Symbols NCAPD3
Host Species Rabbit
Immunogen Recombinant Human NCAPD3. Recombinant protein control fragment (Product #RP-105978).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23310
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.