Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCAPG Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578557
Description
Positive Control: 293T, A431 Predicted Reactivity: Bovine (81%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a subunit of the condensin complex, which is responsible for the condensation and stabilization of chromosomes during mitosis and meiosis. Phosphorylation of the encoded protein activates the condensin complex. There are pseudogenes for this gene on chromosomes 8 and 15. Alternative splicing results in multiple transcript variants.
Specifications
NCAPG | |
Polyclonal | |
Unconjugated | |
NCAPG | |
5730507H05Rik; CAPG; CHCG; chromosome condensation protein G; chromosome-associated protein G; Condensin complex subunit 3; condensin subunit CAP-G; FLJ12450; Hcapg; hCAP-G; Melanoma antigen NY-MEL-3; MFT.M05.13; MGC126525; NCAPG; Non-SMC condensin I complex subunit G; non-SMC condensin I complex, subunit G; NYMEL3; NY-MEL-3; on-SMC condensin I complex, subunit G; RGD1562646; XCAP-G homolog; YCG1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64151 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.89 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9BPX3 | |
NCAPG | |
Recombinant protein encompassing a sequence within the center region of human HCAP-G. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction