Learn More
Invitrogen™ NCAPG2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total NCAPG2 protein.
Specifications
Specifications
| Antigen | NCAPG2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | NCAPG2 |
| Gene Accession No. | Q86XI2 |
| Gene Alias | 5830426I05Rik; CAPG2; CAP-G2; chromosome-associated protein G2; Condensin-2 complex subunit G2; hCAP-G2; leucine zipper protein 5; LUZP5; mCAP-G2; more than blood homolog; more than blood protein; Mtb; Ncapg2; Non-SMC condensin II complex subunit G2; non-SMC condensin II complex, subunit G2; RGD1308406 |
| Gene Symbols | NCAPG2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.