Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCAPG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140072 Supplier Invitrogen™ Supplier No. PA5140072
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DGREKENVTV LDKTLSVNDV ACMAGLLEII VILWKSIDRS MENNKEAKLY TINKFASVLP EYLKVFKDDR CKIPLFMLMS FMPASAVPPF SCGVISTLR Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 85%.

This gene encodes a protein that belongs to the Condensin2nSMC family of proteins. The encoded protein is a regulatory subunit of the condensin II complex which, along with the condensin I complex, plays a role in chromosome assembly and segregation during mitosis. A similar protein in mouse is required for early development of the embryo. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Aug 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAPG2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCAPG2
Gene Accession No. Q86XI2
Gene Alias 5830426I05Rik; CAPG2; CAP-G2; chromosome-associated protein G2; Condensin-2 complex subunit G2; hCAP-G2; leucine zipper protein 5; LUZP5; mCAP-G2; more than blood homolog; more than blood protein; Mtb; Ncapg2; Non-SMC condensin II complex subunit G2; non-SMC condensin II complex, subunit G2; RGD1308406
Gene Symbols NCAPG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NCAPG2. Recombinant protein control fragment (Product #RP-108875).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54892
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.