Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCAPH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528559 Supplier Invitrogen™ Supplier No. PA528559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAPH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene NCAPH
Gene Accession No. Q15003
Gene Alias A730011O11Rik; barren homolog 1; barren homolog protein 1; Brrn; BRRN1; Caph; CAP-H; Chromosome-associated protein H; condensin complex subunit 2; HCAP-H; KIAA0074; mCAP-H; mKIAA0074; Ncaph; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog
Gene Symbols NCAPH
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BRRN1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23397
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.