Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCAPH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580842
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NCAPH.
This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.
Specifications
NCAPH | |
Polyclonal | |
Unconjugated | |
NCAPH | |
A730011O11Rik; barren homolog 1; barren homolog protein 1; Brrn; BRRN1; Caph; CAP-H; Chromosome-associated protein H; condensin complex subunit 2; HCAP-H; KIAA0074; mCAP-H; mKIAA0074; Ncaph; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
23397 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with no preservative | |
Q15003 | |
NCAPH | |
A synthetic peptide corresponding to the C-terminus of the Human NCAPH. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction