Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAPH Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988008
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8008 Supplier Bioss Supplier No. BS7780R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.

Specifications

Antigen NCAPH
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NCAPH
Gene Accession No. Q15003
Gene Alias A730011O11Rik; barren homolog 1; Barren homolog protein 1; BRRN; Brrn1; Caph; CAP-H; Chromosome-associated protein H; Condensin complex subunit 2; hCAP-H; KIAA0074; mCAP-H; mKIAA0074; NCAPH; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog
Gene Symbols NCAPH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NCAPH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23397
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.