Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCAPH2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128723B
Description
NCAPH2 Polyclonal antibody specifically detects NCAPH2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NCAPH2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CAP-H2, CAP-H2 subunit of the condensin II complex, CAPH2,384D8-2, Chromosome-associated protein H2, condensin-2 complex subunit H2, hCAP-H2, kleisin beta, Kleisin-beta, MGC15858, MGC18000, MGC2455, MGC4133, MGC5305, MGC8640, Non-SMC condensin II complex subunit H2, non-SMC condensin II complex, subunit H2 | |
The immunogen recognized by this antibody maps to a region between residue 425 and 475 of human chromosome-associated protein H2 using the numbering given in entry NP_689512.2 (GeneID 29781). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
29781 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction