Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583607 Supplier Invitrogen™ Supplier No. PA583607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVSVTQEEDV PQVRRDWYVY AFLSSLPWVG KELYEKKDAE MDRIFANTES YLKRRQKTHV PMLQVWTADK PHPQEEYLDC LWAQIQKLKK DRWQERHILR PYLAFDSILC EALQHNLPPF TPPPH Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

May be involved in protein trafficking from the plasma membrane to the early endosome (EE) as well as in homotypic fusion of endocytic organelles. Mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCBP1
Gene Accession No. Q09161
Gene Alias 80 kDa nuclear cap-binding protein; AU014645; AW538051; CBP80; MGC2087; NCBP; NCBP 80 kDa subunit; Ncbp1; nuclear cap binding protein subunit 1; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; nuclear cap-binding protein subunit 1; RP11-546O6.1; Sto1
Gene Symbols NCBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NCBP1. Recombinant protein control fragment (Product #RP-91848).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4686
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.