Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115624
Description
Antibody detects endogenous levels of total NCBP1.
May be involved in protein trafficking from the plasma membrane to the early endosome (EE) as well as in homotypic fusion of endocytic organelles. Mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes.
Specifications
NCBP1 | |
Polyclonal | |
Unconjugated | |
NCBP1 | |
80 kDa nuclear cap-binding protein; AU014645; AW538051; CBP80; MGC2087; NCBP; NCBP 80 kDa subunit; Ncbp1; nuclear cap binding protein subunit 1; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; nuclear cap-binding protein subunit 1; RP11-546O6.1; Sto1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
298075, 433702, 4686 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q09161, Q3UYV9, Q56A27 | |
NCBP1 | |
A synthesized peptide derived from human NCBP1(Accession Q09161), corresponding to amino acid residues M1-I35. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction