Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCBP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575234
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5' cap of nascent pre-mRNA in the nucleoplasm. The encoded protein has an RNP domain commonly found in RNA binding proteins, and contains the cap-binding activity. The CBC promotes pre-mRNA splicing, 3'-end processing, RNA nuclear export, and nonsense-mediated mRNA decay. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
NCBP2 | |
Polyclonal | |
Unconjugated | |
NCBP2 | |
20 kDa nuclear cap-binding protein; 20kDa; 5930413E18Rik; AA536802; AI843301; C79367; CBC2; CBP20; cell proliferation-inducing gene 55 protein; NCBP 20 kDa subunit; NCBP interacting protein 1; NCBP2; NCBP-interacting protein 1; NIP1; nuclear cap binding protein subunit 2; nuclear cap binding protein subunit 2, 20kD; nuclear cap binding protein subunit 2, 20kDa; nuclear cap-binding protein subunit 2; phosphatidylinositol glycan anchor biosynthesis, class Z; PIG55; Pigz | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22916, 68092, 689116 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
B1WC40, P52298, Q9CQ49 | |
NCBP2 | |
Synthetic peptide corresponding to amino acids N-terminus of Human NCBP2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction