Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597995 Supplier Invitrogen™ Supplier No. PA597995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCBP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5' cap of nascent pre-mRNA in the nucleoplasm. The encoded protein has an RNP domain commonly found in RNA binding proteins, and contains the cap-binding activity. The CBC promotes pre-mRNA splicing, 3'-end processing, RNA nuclear export, and nonsense-mediated mRNA decay. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCBP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NCBP2
Gene Accession No. P52298, Q9CQ49
Gene Alias 20 kDa nuclear cap-binding protein; 20kDa; 5930413E18Rik; AA536802; AI843301; C79367; CBC2; CBP20; cell proliferation-inducing gene 55 protein; NCBP 20 kDa subunit; NCBP interacting protein 1; NCBP2; NCBP-interacting protein 1; NIP1; nuclear cap binding protein subunit 2; nuclear cap binding protein subunit 2, 20kD; nuclear cap binding protein subunit 2, 20kDa; nuclear cap-binding protein subunit 2; phosphatidylinositol glycan anchor biosynthesis, class Z; PIG55; Pigz
Gene Symbols NCBP2
Host Species Rabbit
Immunogen Recombinant Human Nuclear cap-binding protein subunit 2 protein (1-156AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22916, 68092
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.