Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCBP2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582403
Description
Immunogen sequence: RDHQFSGRKF QQEKLLKESS TLNMGNLSFY TTEEKIHELF SRSDIRNIFM GLDKIKKTAC GFCFVECHNR ADAENAMRFL TGTCLDEWII CTDWDVGFRE GQQ.
NCBP2L is a protein coding gene. Gene ontology (GO) annotation include nuclear cap binding complex.
Specifications
NCBP2L | |
Polyclonal | |
Unconjugated | |
NCBP2L | |
DJ820B18.1; Nuclear Cap Binding Protein Subunit 2 Like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
392517 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6PVI3 | |
NCBP2L | |
Recombinant protein corresponding to Human NCBP2L. Recombinant protein control fragment (Product #RP-92235). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction