Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCCRP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NCCRP1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NCCRP1 Polyclonal specifically detects NCCRP1 in Human samples. It is validated for Western Blot.Specifications
NCCRP1 | |
Polyclonal | |
Rabbit | |
Adaptive Immunity, Immunology | |
Q6ZVX7 | |
342897 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
LOC342897, NCCRP-1, nonspecific cytotoxic cell receptor protein 1 homolog, non-specific cytotoxic cell receptor protein 1 homolog, non-specific cytotoxic cell receptor protein 1 homolog (zebrafish) | |
NCCRP1 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title