Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCDN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544254 Supplier Invitrogen™ Supplier No. PA544254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSCCDLAAAG QLGKASIMAS DCEPALNQAE GRNPTLERYL GALREAKNDS Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.

Probably involved in signal transduction, in the nervous system, via increasing cell surface localization of GRM5 and positively regulating its signaling. Required for the spatial learning process. Acts as a negative regulator of Ca(2+)-calmodulin-dependent protein kinase 2 (CaMK2) phosphorylation. May play a role in modulating melanin-concentrating hormone-mediated functions via its interaction with MCHR1 that interferes with G protein-coupled signal transduction. May be involved in bone metabolism. May also be involved in neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCDN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NCDN
Gene Accession No. Q9UBB6
Gene Alias AU042419; Kiaa0607; minisatellite 10ae detected by probe MMS10; mKIAA0607; MMS10-AE; Ms10ae; M-Sema F-associating protein of 75 kDa; NCDN; Neurite outgrowth-related protein from the rat brain; neurochondrin; neurochondrin-1; neurochondrin-2; Norbin; Sfap75; similar to neurochondrin
Gene Symbols NCDN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NCDN (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23154
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.