Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCE2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142723
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
Accepts the ubiquitin-like protein NEDD8 from the UBA3-NAE1 E1 complex and catalyzes its covalent attachment to other proteins. The specific interaction with the E3 ubiquitin ligase RBX2, but not RBX1, suggests that the RBX2-UBE2F complex neddylates specific target proteins, such as CUL5.
Specifications
NCE2 | |
Polyclonal | |
Unconjugated | |
UBE2F | |
2510010F15Rik; AI851109; MGC18120; NCE2; NEDD8 carrier protein UBE2F; NEDD8 protein ligase UBE2F; NEDD8-conjugating enzyme 2; NEDD8-conjugating enzyme UBE2F; RGD1307608; RING-type E3 NEDD8 transferase UBE2F; UBE2F; Ube2fa; ubiquitin conjugating enzyme E2 F (putative); ubiquitin conjugating enzyme E2F (putative); ubiquitin-conjugating enzyme E2 F; ubiquitin-conjugating enzyme E2F (putative) | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
363284 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q5U203 | |
UBE2F | |
Peptide with sequence SKLKRDDGLKGSRT-C. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction