Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCE2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988511
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8511 Supplier Bioss Supplier No. BS8378R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Accepts the ubiquitin-like protein NEDD8 from the UBA3-NAE1 E1 complex and catalyzes its covalent attachment to other proteins. The specific interaction with the E3 ubiquitin ligase RBX2, but not RBX1, suggests that the RBX2-UBE2F complex neddylates specific target proteins, such as CUL5.

Specifications

Antigen NCE2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UBE2F
Gene Accession No. Q969M7
Gene Alias 2510010F15Rik; AI851109; MGC18120; NCE2; NEDD8 carrier protein UBE2F; NEDD8 protein ligase UBE2F; NEDD8-conjugating enzyme 2; NEDD8-conjugating enzyme UBE2F; RGD1307608; RING-type E3 NEDD8 transferase UBE2F; UBE2F; Ube2fa; ubiquitin conjugating enzyme E2 F (putative); ubiquitin conjugating enzyme E2F (putative); Ubiquitin-conjugating enzyme E2 F; ubiquitin-conjugating enzyme E2F (putative)
Gene Symbols UBE2F
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NCE2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140739
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.