Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCEH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550285 Supplier Invitrogen™ Supplier No. PA550285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total NCEH1 protein.

Neutral cholesterol ester hydrolase 1 (NCEH) also known as arylacetamide deacetylase-like 1 (AADACL1) or KIAA1363 is an enzyme that in humans is encoded by the NCEH1 gene. NCEH is an enzyme located in the endoplasmic reticulum. NCEH hydrolyzes 2-acetyl monoalkylglycerol ether, as part of an enzymatic pathway regulating the levels of platelet activating factor and lysophospholipids that may be involved in cancer development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCEH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NCEH1
Gene Accession No. Q6PIU2, Q8BLF1
Gene Alias AADACL1; Arylacetamide deacetylase-like 1; B230106I24Rik; chlorpyrifos oxon-binding protein; CPO-BP; Kiaa1363; NCEH; NCEH1; Neutral cholesterol ester hydrolase 1; RGD1311104
Gene Symbols NCEH1
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human Neutral cholesterol ester hydrolase 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320024, 57552
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.