Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCEH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555453
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555453 Supplier Invitrogen™ Supplier No. PA555453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALIYPVLQAL DFNTPSYQQN VNTPILPRYV MVKYWVDYFK GNYDFVQAMI VNNHTSLDVE EAAAVRARLN WTSLLPASFT KNYKPVVQTT GNARIVQELP QLLDARSAPL IADQAVLQLL PKTYILTCEH DVL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

Neutral cholesterol ester hydrolase 1 (NCEH) also known as arylacetamide deacetylase-like 1 (AADACL1) or KIAA1363 is an enzyme that in humans is encoded by the NCEH1 gene. NCEH is an enzyme located in the endoplasmic reticulum. NCEH hydrolyzes 2-acetyl monoalkylglycerol ether, as part of an enzymatic pathway regulating the levels of platelet activating factor and lysophospholipids that may be involved in cancer development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCEH1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCEH1
Gene Accession No. Q6PIU2
Gene Alias AADACL1; Arylacetamide deacetylase-like 1; B230106I24Rik; chlorpyrifos oxon-binding protein; CPO-BP; Kiaa1363; NCEH; NCEH1; Neutral cholesterol ester hydrolase 1; RGD1311104
Gene Symbols NCEH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NCEH1. Recombinant protein control fragment (Product #RP-90318).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.