Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCF1 Antibody (SR1776), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322192100UL
Description
NCF1 Monoclonal antibody specifically detects NCF1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
NCF1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
SR1776 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
47 kDa neutrophil oxidase factor, FLJ79451, NADPH oxidase organizer 2, NCF-1, NCF1A, neutrophil cytosol factor 1,47 kDa autosomal chronic granulomatous disease protein, neutrophil cytosolic factor 1, neutrophil cytosolic factor 1 (47kD, chronic granulomatous disease, autosomal1), neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1), Neutrophil NADPH oxidase factor 1, Nox organizer 2, NOXO2NCF-47K, Nox-organizing protein 2, p47phox, SH3 and PX domain-containing protein 1A, SH3PXD1Ap47-phox | |
A synthesized peptide derived from human NCF1 (Uniprot #: P14598) | |
100 μg | |
Cancer, Endocrinology | |
653361 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction