Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCF1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549625
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549625 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549625 Supplier Invitrogen™ Supplier No. PA549625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCF1C Polyclonal Antibody for Western Blot, IHC (P), Flow

May be required for activation of the latent NADPH oxidase (necessary for superoxide production).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCF1C
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NCF1C
Gene Accession No. A8MVU1
Gene Alias NCF1C; NCF-1C; neutrophil cytosolic factor 1C pseudogene; SH3PXD1C
Gene Symbols NCF1C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 277-303 amino acids from the C-terminal region of human NCF1C.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 654817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.