Learn More
Invitrogen™ NCF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551702
Description
Immunogen sequence: QEESSPQSDI PAPPSSKAPG RPQLSPGQKQ KEEPKEVKLS VPMPYTLKVH YKYTVVMKTQ PGLPYSQVRD MVSKKLELRL EHTKLSYRPR DSNELVPLSE DSMKDAWGQV KNYCLTLWCE NTVGDQGFPD EPKESEKADA NNQTTEPQ Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 24%.
NCF2, NCF1, and a membrane bound cytochrome b558 are required for activation of the latent NADPH oxidase (necessary for superoxide production). Defects in NCF2 are a cause of chronic granulomatous disease autosomal recessive cytochrome-b-positive type 2 (CGD2). Chronic granulomatous disease is a genetically heterogeneous disorder characterized by the inability of neutrophils and phagocytes to kill microbes that they have ingested. Patients suffer from life-threatening bacterial/fungal infections.
Specifications
NCF2 | |
Polyclonal | |
Unconjugated | |
NCF2 | |
67 kDa neutrophil oxidase factor; chronic granulomatous disease, autosomal 2; EMBL:FAA00361.1}; FLJ93058; NADPH oxidase activator 2; NADPH oxidase subunit (67 kD); NADPH oxidase subunit (67kDa); NCF2; NCF-2; ncf2 {ECO:0000312; neutrophil cytosol factor 2; neutrophil cytosolic factor 2; neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal 2); neutrophil NADPH oxidase factor 2; Noxa2; p67phox; p67-phox | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4688 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P19878 | |
NCF2 | |
Recombinant protein corresponding to Human NCF2. Recombinant protein control fragment (Product #RP-95408). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.