Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597997
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597997 Supplier Thermo Scientific Supplier No. PA597997
Add to Cart
Add to Cart

NCF2 Polyclonal Antibody

NCF2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen NCF2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NCF2
Gene Accession No. P19878
Gene Alias 67 kDa neutrophil oxidase factor; chronic granulomatous disease, autosomal 2; EMBL:FAA00361.1}; FLJ93058; NADPH oxidase activator 2; NADPH oxidase subunit (67 kD); NADPH oxidase subunit (67kDa); NCF2; NCF-2; ncf2 {ECO:0000312; neutrophil cytosol factor 2; neutrophil cytosolic factor 2; neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal 2); neutrophil NADPH oxidase factor 2; Noxa2; p67phox; p67-phox
Gene Symbols NCF2
Host Species Rabbit
Immunogen Recombinant Human Neutrophil cytosol factor 2 protein (178-526AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4688
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.