Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCF4 Rabbit anti-Human, Mouse, Clone: 4L9Z4, Novus Biologicals™
SDP

Catalog No. p-7246860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165741 20 μg
NB165740 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165741 Supplier Novus Biologicals Supplier No. NBP31532220UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NCF4 Monoclonal antibody specifically detects NCF4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen NCF4
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4L9Z4
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias MGC3810, NCF, NCF-4, neutrophil cytosol factor 4, neutrophil cytosolic factor 4 (40kD), neutrophil cytosolic factor 4, 40kDa, Neutrophil NADPH oxidase factor 4, p40phox, p40-phox, SH3 and PX domain-containing protein 4, SH3PXD4P40PHOX
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 240-339 of human NCF4 (Q15080).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4689
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.