Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nck1/NCK alpha Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983319
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3319 Supplier Bioss Supplier No. BS1466R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.

Specifications

Antigen Nck1/NCK alpha
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Nck1
Gene Accession No. P16333
Gene Alias 6330586M15Rik; Cytoplasmic protein NCK1; D230010O13Rik; melanoma NCK protein; MGC12668; NCK; NCK adaptor protein 1; NCK tyrosine kinase; Nck1; Nck-1; NCKalpha; non-catalytic region of tyrosine kinase; non-catalytic region of tyrosine kinase adaptor protein 1; SH2/SH3 adaptor protein NCK-alpha
Gene Symbols Nck1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Nck2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4690
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.