Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102331 Supplier Invitrogen™ Supplier No. PA5102331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NCK1.

NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCK1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Nck1
Gene Accession No. P16333, Q99M51
Gene Alias 6330586M15Rik; Cytoplasmic protein NCK1; D230010O13Rik; melanoma NCK protein; MGC12668; Nck; NCK adaptor protein 1; NCK tyrosine kinase; NCK1; nck-1; NCKalpha; non-catalytic region of tyrosine kinase; non-catalytic region of tyrosine kinase adaptor protein 1; SH2/SH3 adaptor protein NCK-alpha
Gene Symbols Nck1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NCK1(Accession P16333), corresponding to amino acid residues S66-N116.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17973, 300955, 4690
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.