Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCK1 Recombinant Rabbit Monoclonal Antibody (JU41-80)
GREENER_CHOICE

Catalog No. PIMA534616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534616 Supplier Invitrogen™ Supplier No. MA534616
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: NIH-3T3, Hela, rat kidney tissue lysate, mouse testis tissue lysate, A431, HUVEC, LOVO, rat testis tissue, human tonsil tissue, human kidney tissue.

NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCK1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JU41-80
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Nck1
Gene Accession No. P16333
Gene Alias 6330586M15Rik; Cytoplasmic protein NCK1; D230010O13Rik; melanoma NCK protein; MGC12668; Nck; NCK adaptor protein 1; NCK tyrosine kinase; NCK1; nck-1; NCKalpha; non-catalytic region of tyrosine kinase; non-catalytic region of tyrosine kinase adaptor protein 1; SH2/SH3 adaptor protein NCK-alpha
Gene Symbols Nck1
Host Species Rabbit
Immunogen Recombinant protein within Human NCK1 aa 204-296.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4690
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.