Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112625
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112625 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112625 Supplier Invitrogen™ Supplier No. PA5112625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCK2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The NCK family of SH2/SH3 adaptor proteins consists of two members, NCK1 and NCK2, which couple tyrosine kinase signaling, including the EGF and PDGF receptor-pathways, to downstream signaling proteins. The NCK2 protein binds human PDGFR-b (Tyr-1009); overexpression of NCK2 inhibits PDGF-induced membrane ruffling and lamellipod formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCK2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NCK2
Gene Accession No. O43639, O55033
Gene Alias 4833426I10Rik; cytoplasmic protein NCK2; Grb4; growth factor receptor-bound protein 4; NCK adaptor protein 2; NCK2; Nck-2; NCKbeta; non-catalytic region of tyrosine kinase adaptor protein 2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta
Gene Symbols NCK2
Host Species Rabbit
Immunogen Recombinant Human Cytoplasmic protein NCK2 protein (1-380AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17974, 8440
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.